missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) oxide, Puriss., meets analytical spec. of BP, Ph. Eur., USP, 99 to 100.5%, Honeywell Fluka™
BIOPHARMA

Puriss., meets analytical specification of BP, Ph. Eur., USP, 99-100.5%

43.31 € - 705.58 €

Chemical Identifiers

CAS 13463-67-7
Molecular Formula O2Ti
Molecular Weight (g/mol) 79.87
MDL Number MFCD00011269 MFCD00210650
InChI Key GWEVSGVZZGPLCZ-UHFFFAOYSA-N
Synonym titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
PubChem CID 26042
ChEBI CHEBI:32234
IUPAC Name dioxotitanium
SMILES O=[Ti]=O
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15641390
BIOPHARMA
View Documents
Honeywell-Fluka
14027-250G
250 g
43.31 €
250g
Please to purchase this item. Need a web account? Register with us today!
15631390
BIOPHARMA
View Documents
Honeywell-Fluka
14027-1KG
1 kg
102.18 €
1kg
Please to purchase this item. Need a web account? Register with us today!
15661390
View Documents
Honeywell-Fluka
14027-6X1KG
6 x 1 kg
482.81 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15651390
BIOPHARMA
View Documents
Honeywell-Fluka
14027-25KG
25 kg
705.58 €
25kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

13463-67-7
79.87
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
26042
dioxotitanium
O2Ti
MFCD00011269 MFCD00210650
titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
CHEBI:32234
O=[Ti]=O

Specifications

13463-67-7
TiO2
NONH for all modes of transport
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
dioxotitanium
26042
79.87g/mol
puriss.
Titanium(Iv) Oxide, Meets analytical specification of Ph. Eur.BP, USP
O2Ti
MFCD00011269 MFCD00210650
titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
O=[Ti]=O
79.87
CHEBI:32234
99 to 100.5%
4.26g/mL at 25°C
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 236-675-5

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.