Learn More
MP Biomedicals™ Titertek™ Finnpipette™ Multistepper Pipet Tips
Hydrophobic tips with very low coefficient of friction
Brand:  MP Biomedicals™ 0977944H2
188.10 EUR valid until 2025-12-31
Use promo code "24285" to get your promotional price.
 View more versions of this product
Product Code. 11409625
  • 198.00 € / Pack of 960
Estimated Shipment: 02-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Specifically for the Titertek™/Labsystems 8-channel Multistepper pipette
  • Made from virgin polypropylene material
  • Comes with robot-compatible (large-micron) hydrophobic filters to eliminate instrument contamination
  • 96 tips per rack
  • Volume: 1500μL
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1500 μL
Natural
Polypropylene
Sterile
Rack