missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Titertek™ Finnpipette™ Multistepper Pipet Tips

Hydrophobic tips with very low coefficient of friction

148.00 € - 198.00 €

Specifications

Volume (Metric) 1500 μL
Color Natural
Material Polypropylene
View More Specs

Products 2
Product Code Brand Sterility Format Price Quantity & Availability  
Product Code Brand Sterility Format Price Quantity & Availability  
11489615
View Documents
MP Biomedicals™
0977943C2
Non-sterile Bulk
148.00 €
Pack of 1000
Estimated Shipment: 20-03-2025
to see stock.
Add to basket
 
11409625
View Documents
MP Biomedicals™
0977944H2
Sterile Rack
198.00 €
Pack of 960
Estimated Shipment: 20-03-2025
to see stock.
Add to basket
 
Description

Description

Specifically for the Titertek™/Labsystems 8-channel Multistepper pipette

 

  • Made from virgin polypropylene material
  • Comes with robot-compatible (large-micron) hydrophobic filters to eliminate instrument contamination
  • 96 tips per rack
  • Volume: 1500μL

 

Specifications

Specifications

1500 μL
Natural
Polypropylene
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.