Learn More
Tuttnauer Tiva8 Glass Washer Disinfector
The Tiva8 Glass Washer Disinfector provides a flexible turnkey solution for washing, disinfecting and drying diverse laboratory equipment for a complete decontamination life cycle. The washer can be loaded up to 4 washing levels with a large variety of washing trolleys and accessories.
Brand:  Tuttnauer TIVA8-WD-1ML
 View more versions of this product
Product Code. 17910050
  • 12030.00 € / 1 unit
Explore more special offers
Description
Description
Small, reliable and customizable. The Tiva 8 washer disinfector provides a flexible turnkey solution for washing, disinfecting and drying diverse laboratory equipment for a complete decontamination life cycle. The TIVA 8 washer can be loaded up to 4 washing levels with a large variety of washing trolleys and accessories.
  • Diverse load types
    • Available trolleys for DIN baskets, containers, mini-invasive equipment, anesthesia kits, bottles and shoes
  • Washing chamber made of high quality AISI 316L
    • 165 Liter chamber, perfect for private clinics
  • Manual glass door
    • Hinged, manual door
  • Advanced Touch Control Touch panel with color graphic display with a total of 40 programs: preset and programmable procedures
Advanced features
  • Air drying prefilter and HEPA H14 filter.
  • 3 heating elements.
  • Built-in steam condenser.
  • Integrated water softener (optional).
  • Dosing pumps with flowmeter
  • For acid and alkaline detergents
  • 2 additional pumps optional
Specifications
Specifications
Wide with Glass Door and Side Cabinet
Stainless Steel
43.6 gal.
LCD
Laboratory Glassware Washer
165 L
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products