Learn More
Thermo Scientific Chemicals TLC Grade 12 Micron APS Silica Gel
Brand:  Thermo Scientific Chemicals 042765.18
 View more versions of this product
Product Code. 11305587
  • 20.60 € / 50g
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
TLC plate preparation
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
12 μm
TLC
50 g
Silica Gel
60 Å
6.5 to 7.5
500 to 600 m2/g
Silica