missing translation for 'onlineSavingsMsg'
Learn More

Toluene, 99+%, extra pure, Thermo Scientific Chemicals

42.81 € - 579.89 €

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10702991
View Documents
Thermo Scientific Acros
177160010
1 L Glass Bottle
42.81 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10712991
View Documents
Thermo Scientific Acros
177160025
2.5 L Glass Bottle
80.80 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10417153
View Documents
Thermo Scientific Acros
177160100
10 L Plastic Drum
257.28 €
10 L
Please to purchase this item. Need a web account? Register with us today!
10367210
View Documents
Thermo Scientific Acros
177160250
25 L Metal Drum
579.89 €
25 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

108-88-3
92.14
YXFVVABEGXRONW-UHFFFAOYSA-N
1140
toluene
C7H8
MFCD00008512
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
CHEBI:17578
CC1=CC=CC=C1

Specifications

-95.0°C
0.8660g/mL
4°C
1.4968
0.003% max.
C7H8
MFCD00008512
0.6 mPa.s (20°C)
0.05% max. (K.F.)
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
YXFVVABEGXRONW-UHFFFAOYSA-N
toluene
1140
92.14
Extra Pure
108-88-3
111.0°C
Liquid
99+%
0.866
C6H5CH3
Vapor Pressure: 29mbar at 20°C
05, 280
15, 9688
Solubility in water: almost insoluble (0.5g/L at 20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol, acetone,, ether, petroleum ether, benzene, ch2cl2, dmf, gla, cial acetic acid, chcl3, ccl4.
CC1=CC=CC=C1
92.14
CHEBI:17578
≥99%
Toluene
Videos
Safety and Handling

Safety and Handling

GHS P Statement May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure if inhaled. May be fatal if swallowed and enters airways. Causes skin irritation. Suspected of damag

Emergency Overview
Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects.
NFPA
Health:2 Flammability:3 Instability:0

Signal Word: DANGER!

Product Identifier
  • Toluene

Signal Word
  • Danger

Hazard Category
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only