Learn More
Topiramate, MedChemExpress
MedChemExpress Topiramate (McN 4853) is a broad-spectrum antiepileptic agent. Topiramate is a GluR5 receptor antagonist. Topiramate produces its antiepileptic effects through enhancement of GABAergic activity, inhibition of kainate/AMPA receptors, inhibition of voltage-sensitive sodium and calcium channels, increases in potassium conductance, and inhibition of carbonic anhydrase.
130.95 € - 394.20 €
Specifications
Chemical Name or Material | Topiramate |
---|---|
CAS | 97240-79-4 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C12H21NO8S |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16492026
|
Medchem Express
HY-B0122 |
500 mg |
394.20 €
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16458116
|
Medchem Express
HY-B0122 |
100 mg |
130.95 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Topiramate (McN 4853) is a broad-spectrum antiepileptic agent. Topiramate is a GluR5 receptor antagonist. Topiramate produces its antiepileptic effects through enhancement of GABAergic activity, inhibition of kainate/AMPA receptors, inhibition of voltage-sensitive sodium and calcium channels, increases in potassium conductance, and inhibition of carbonic anhydrase.Specifications
Topiramate | |
White | |
C12H21NO8S | |
DMSO : ≥ 100 mg/mL (294.67 mM) ∣H2O : 4 mg/mL (11.79 mM; Need ultrasonic) | |
339.36 | |
98.0% | |
Solid |
97240-79-4 | |
Research | |
McN 4853 RWJ 17021 | |
NS(OC[C@]1(OC(C)(C)O2)[C@@H]2[C@H](OC(C)(C)O3)[C@H]3CO1)(=O)=O | |
339.36 | |
Research | |
Neuroscience-Neuromodulation |
Safety and Handling
H315∣H319∣H335
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only