Learn More
Toray Carbon Paper, PTFE treated, TGP-H-60, 19x19cm, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 045365.KS
171.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Toray Carbon Paper, PTFE treated, TGP-H-60 is used as a gas diffusion layer in fuel cell applications. It is suitable for use as a catalyst backing layer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Toray Carbon Paper | |
Cardboard Box | |
MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731 | |
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
carbon | |
5462310 |
7782-42-5 | |
C | |
1 Ea. | |
Insoluble in water. | |
[C] | |
12.01 | |
CHEBI:27594 |
Product Suggestions
Customers who viewed this item also viewed.