Learn More
Total Base Number Standard, ASTM D 2896 (nominal value: 0.99 mg KOH/g), 1 x 125g, Paragon Scientific™
Specifically manufactured for the verification of analytical instruments used to determine base number by potentiometric titration. Each standard is certified in strict accordance with international test method protocols, ASTM D2896 / IP 276.
Brand:  Paragon Scientific TBN1
Product Code. 16040440
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Base Number (TBN) tested and certified in accordance with ASTM D2896 / IP 276
  • Certified in strict accordance to ISO 17025 and ISO 17034 under our UKAS accreditation
  • Highest level of accreditation guarantee, providing the most credible certified data available worldwide
  • Fully traceable to international standards and low levels of uncertainty achieves maximum accuracy of data
  • Typically used in, but not limited to, the analysis of used oils and lubricants
  • Supplied as single volume packs or as triple packs
  • Supplied in tamper-evident glass packaging, providing assurance of sample integrity
  • Manufactured in the United Kingdom
  • 24 month shelf life
Specifications
Specifications
1 x 125 g
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Total Base Number Standard, ASTM D 2896 (nominal value: 0.99 mg KOH/g), 1 x 125g, Paragon Scientific™ > Quantity: 1 x 125g
Nominal values quoted. Certified Values will vary from batch to batch.