missing translation for 'onlineSavingsMsg'
Learn More

Total Organic Carbon (TOC), standard solution, Specpure™ 1000μg/mL, Thermo Scientific Chemicals

141.71 € - 282.41 €

Specifications

Chemical Name or Material Total Organic Carbon
Name Note 1000μg/mL
Color Colorless
Molecular Formula KHC8H4O4 in H2O
MDL Number MFCD00133992
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11329168
View Documents
Thermo Scientific Alfa Aesar
042562.AE
100 mL
141.71 €
100mL
Please to purchase this item. Need a web account? Register with us today!
11366109
View Documents
Thermo Scientific Alfa Aesar
042562.AP
500 mL
282.41 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Total Organic Carbon (TOC), is used as a standard solution during the measurement of chemical analysis such as detecting contaminants in drinking water and cooling water. It is also used for analysis of water to be used for semiconductor manufacturing and pharmaceutical use.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Total Organic Carbon
Colorless
MFCD00133992
Miscible with water
1000μg/mL
KHC8H4O4 in H2O
Characteristic
Liquid
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only