Learn More
Bellingham and Stanley™ Touch Screen Refractometer
Peltier temperature controlled refractometer with resolution touch display
Brand:  Bellingham and Stanley™ 19-40
 View more versions of this product
Product Code. 15807205
  • 12240.00 € / Each
Estimated Shipment: 24-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Touch Screen Refractometer with Peltier temperature control features the latest integral 7 in. full color touchscreen, high definition display, and USB/LAN connectivity. The refractometer is supplied with two RFID tags, instruction manual, and certificate of conformity. Perfect for measurement and control in demanding food, beverage, chemical and industrial applications.
  • Scales
    • RI 1.32 - 1.58
    • Brix 0 - 100
  • Resolution:
    • RI 0.00001 (RFM330-T)/ 0.000001 (RFM340-T)
    • Brix 0.01
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Digital Refractometer
Resolution: 0.01/0.001 Brix
± 0.01 to 0.03 Brix
0 to 100 Brix
RFM340-T
Refractometer, RFID Tags (2), Instruction Manual, and Certificate of Conformity
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Bellingham and Stanley™ Touch Screen Refractometer > 0.01/0.001 Brix