missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell TouchNTuff™ 93-163 Series Blue Disposable Nitrile Gloves
Proven splash resistance against hazardous chemicals. Ansell TouchNTuff™ 93-163 Series Blue Disposable Nitrile Gloves are flexible and comfortable offering better dexterity.
38.75 €
Specifications
Certifications/Compliance | EN 420:2003 + A1:2009 and EN 374:2003 |
---|---|
Color | Blue |
Disposable | Single-use |
Exterior Finish | Fully Textured |
Material | Nitrile |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15532505
|
Ansell
93163110 |
10.5/11 |
38.75 €
Pack of 50 |
Estimated Shipment: 10-01-2025
Log in to see stock. |
|||||
15542505
|
Ansell
93163080 |
7.5/8 |
38.75 €
Pack of 50 |
Estimated Shipment: 10-01-2025
Log in to see stock. |
|||||
15552505
|
Ansell
93163090 |
8.5/9 |
38.75 €
Pack of 50 |
In Stock
Log in to see stock. |
|||||
15522505
|
Ansell
93163100 |
9.5/10 |
38.75 €
Pack of 50 |
In Stock
Log in to see stock. |
|||||
15511265
|
Ansell
93163070 |
6.5/7 |
38.75 €
Pack of 50 |
In Stock
Log in to see stock. |
|||||
Description
- Single use gloves
- Nitrile formulation for low hand fatigue
- Easy donning
- Textured finish, light and resistant
- Good resistance to puncture
- Flexible and comfortable offering excellent dexterity
- Beaded cuff for an easy donning and offering good tear resistance
- Ambidextrous
Applications:
- Chemical handling
- Glass manufacturing
- Handling of cytostatics
- Intricate parts handling
- Laboratory analysis
- Light assembly of oil-coated pieces
- Light assembly tasks
- Paint and spray shops
- Pharmaceuticals
- Printing industry
Specifications
EN 420:2003 + A1:2009 and EN 374:2003 | |
Single-use | |
Nitrile | |
Nitrile Gloves | |
Beaded/Rolled | |
93-163 |
Blue | |
Fully Textured | |
Powder Free | |
0.17 mm | |
50 gloves in a box, 10 boxes in a carton | |
355 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title