Learn More
Trace Metals 1 without Mercury For Environmental Analysis, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
236.00 € - 351.00 €
Specifications
Chemical Name or Material | Trace Metals I without Mercury |
---|---|
Name Note | Groundwater and Wastewater Pollution Control Check Standard |
Packaging | Triple-leached LDPE bottle |
Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15216346
|
SPEX Certiprep
WP-15N |
125 mL | Triple-leached LDPE bottle |
236.00 €
125mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15206346
|
SPEX Certiprep
WP-15-500N |
500 mL | Triple-leached LDPE bottle |
351.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Al | 500μg/mL |
V | 250μg/mL |
As | 100μg/mL |
Be | 100μg/mL |
Co | 100μg/mL |
Cr | 100μg/mL |
Cu | 100μg/mL |
Fe | 100μg/mL |
Mn | 100μg/mL |
Ni | 100μg/mL |
Pb | 100μg/mL |
Zn | 100μg/mL |
Cd | 25μg/mL |
Se | 25μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Trace Metals I without Mercury | |
Triple-leached LDPE bottle | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
Various |
Groundwater and Wastewater Pollution Control Check Standard | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Environmental | |
5% HNO3 |
Safety and Handling
Warning: Causes skin irritation, serious eye irritation Wear protective gloves/eye/face protection. If in eyes, rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If skin/eye irritation occurs, get medical advice/attention.
missing translation for 'dotInformation' : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.