Learn More
Trace Metals 1 without Mercury For Environmental Analysis, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep WP-15N
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Al | 500μg/mL |
V | 250μg/mL |
As | 100μg/mL |
Be | 100μg/mL |
Co | 100μg/mL |
Cr | 100μg/mL |
Cu | 100μg/mL |
Fe | 100μg/mL |
Mn | 100μg/mL |
Ni | 100μg/mL |
Pb | 100μg/mL |
Zn | 100μg/mL |
Cd | 25μg/mL |
Se | 25μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Trace Metals I without Mercury | |
Triple-leached LDPE bottle | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
5% HNO3 | |
Various |
Groundwater and Wastewater Pollution Control Check Standard | |
125 mL | |
Wastewater Testing | |
Certified Reference Material | |
Environmental |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product intended for laboratory use only