missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Trace Minerals for an Ultra-Clean Environment
Trace Minerals is used in ultra clean environments where trace elements usually present, are absent.
Brand: MP Biomedicals™ 0296026401
This item is not returnable.
View return policy
Description
- 7-trace elements which are usually present in a open environment
- Research animal diet supplement
- Powdered

Specifications
Animal Diet | |
Powder | |
1 kg | |
15°C to 30°C |
Silicon Dioxide 10.7%, Nickel Chloride 0.067%, Sodium Fluoride 0.060%, Cobalt Chloride 0.070%, Ammonium Molybdate 0.013%, Tin (Metal) 0.010%, Vanadium Sulfate.2H2O 0.004%, Alphacel, Non-Nutritive Bulk 89.076% | |
Trace Mineral is used in ultra clean environments where trace elements usually present, are absent | |
Rat |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Trace Minerals for an Ultra Clean Environment
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 1B
- Reproductive toxicity Category 1
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H350-May cause cancer.
- H360-May damage fertility or the unborn child.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P321-Specific treatment (see on this label).
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Alphacel, Cobalt Chloride Hexahydrate, nickel dichloride
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MP Biomedicals™ Trace Minerals for an Ultra-Clean Environment > Trace Minerals for an Ultra-Clean Environment, 1 kg
Spot an opportunity for improvement?Share a Content Correction