missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Trace Minerals for an Ultra-Clean Environment
Trace Minerals is used in ultra clean environments where trace elements usually present, are absent.
287.26 €
Description
- 7-trace elements which are usually present in a open environment
- Research animal diet supplement
- Powdered
Specifications
Animal Diet |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Trace Minerals for an Ultra Clean Environment
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 1B
- Reproductive toxicity Category 1
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H350-May cause cancer.
- H360-May damage fertility or the unborn child.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P321-Specific treatment (see on this label).
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Alphacel, Cobalt Chloride Hexahydrate, nickel dichloride