Learn More
Thermo Scientific™ TRACE™ TR-5MS GC Columns
Choose these colums that feature a popular GCMS phase for many applications.
357.00 € - 1360.00 €
Specifications
Stationary Phase Trace™ TR-5 MS
USP Type G27, G36
Polarity Non-Polar
View More Specs
Products 23
Product Code Brand Length (Metric) Diameter (Metric) Inner Film Thickness Price Quantity & Availability  
Product Code Brand Length (Metric) Diameter (Metric) Inner Film Thickness Price Quantity & Availability  
10239694
View Documents
Thermo Scientific™
260F020P
10 m 0.1 mm 0.1 μm
450.02 €
Each
Estimated Shipment: 15-04-2025
to see stock.
Add to basket
10557265
View Documents
Thermo Scientific™
260F024P
10 m 0.32 mm 0.1 μm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10331265
View Documents
Thermo Scientific™
260F035P
15 m 0.25 mm 0.1 μm
390.14 €
Each
Estimated Shipment: 17-04-2025
to see stock.
Add to basket
10197564
View Documents
Thermo Scientific™
260F047P
30 m 0.25 mm 0.1 μm
708.82 €
Each
Estimated Shipment: 22-04-2025
to see stock.
Add to basket
10434403
View Documents
Thermo Scientific™
260F130P
15 m 0.25 mm 0.25 μm
395.79 €
Each
In Stock
to see stock.
Add to basket
10533795
View Documents
Thermo Scientific™
260F142P
30 m 0.25 mm 0.25 μm
680.75 €
Each
Estimated Shipment: 01-05-2025
to see stock.
Add to basket
10592065
View Documents
Thermo Scientific™
260F143P
30 m 0.32 mm 0.25 μm
736.16 €
Each
Estimated Shipment: 06-05-2025
to see stock.
Add to basket
10029382
View Documents
Thermo Scientific™
260F154P
60 m 0.25 mm 0.25 μm
1168.13 €
Each
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
10486954
View Documents
Thermo Scientific™
260F213P
15 m 0.53 mm 0.5 μm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10089422
View Documents
Thermo Scientific™
260F223P
30 m 0.25 mm 0.5 μm
680.75 €
Each
Estimated Shipment: 24-04-2025
to see stock.
Add to basket
10650174
View Documents
Thermo Scientific™
260F224P
30 m 0.32 mm 0.5 μm
767.22 €
Each
Estimated Shipment: 18-04-2025
to see stock.
Add to basket
10380975
View Documents
Thermo Scientific™
260F225P
30 m 0.53 mm 0.5 μm
791.26 €
Each
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
10240105
View Documents
Thermo Scientific™
260F242P
100 m 0.32 mm 0.5 μm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10253553
View Documents
Thermo Scientific™
260F284P
15 m 0.25 mm 1 μm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10038952
View Documents
Thermo Scientific™
260F285P
15 m 0.32 mm 1 μm
416.14 €
Each
Estimated Shipment: 25-04-2025
to see stock.
Add to basket
10495533
View Documents
Thermo Scientific™
260F296P
30 m 0.25 mm 1 μm
699.98 €
Each
Estimated Shipment: 14-04-2025
to see stock.
Add to basket
10593025
View Documents
Thermo Scientific™
260F297P
30 m 0.32 mm 1 μm
757.65 €
Each
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
10659475
View Documents
Thermo Scientific™
260F298P
30 m 0.53 mm 1 μm
827.90 €
Each
Estimated Shipment: 22-04-2025
to see stock.
Add to basket
10753867
View Documents
Thermo Scientific™
260F308P
60 m 0.25 mm 1 μm
1168.13 €
Each
Estimated Shipment: 23-04-2025
to see stock.
Add to basket
10429054
View Documents
Thermo Scientific™
260F309P
60 m 0.32 mm 1 μm
1259.73 €
Each
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
10722927
View Documents
Thermo Scientific™
260F336P
30 m 0.53 mm 1.5 μm
814.15 €
Each
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
10177934
View Documents
Thermo Scientific™
260F396P
30 m 0.53 mm 3 μm
803.85 €
Each
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
10638895
View Documents
Thermo Scientific™
260F578P
20 m 0.18 mm 0.18 μm
599.33 €
Each
Estimated Shipment: 24-04-2025
to see stock.
Add to basket
Description
Description
Thermo Scientific™ TRACE TR-5MS GC Columns feature a popular GCMS phase for many applications.
  • Nonpolar phase, 5% phenyl polysilphenylene-siloxane
  • Low-bleed and high-stability
  • High signal-to-noise for increased sensitivity
  • High robustness to oxygen and water contamination
  • Similar to B-5, DB-5MS, DB-5.625 XTI-5, BPX5, Rtx-5MS, Rtx- 5, SilMS, AT-5, AT-5MS, 007-5MS, SPB-5, CP-Sil 8CB, Ultra-2, HP-5, HP-5MS, HP5-TA, SPB-5, MDN-5S, VF-5ms, RSL-200, CB-5, OV-5, PE-5, 007-2(MP-5), SE-52, SE-54, PTE-5, CC-5, RH-5ms, ZB-5
Recommended for:
  • Hydrocarbons
  • Solvents
  • Pesticides
  • Herbicides
  • Phenols
  • Amines
Specifications
Specifications
Trace™ TR-5 MS
G27, G36
Non-Polar
Videos
SDS
Documents
Documents
Product Certifications
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Thermo Scientific™ TRACE™ TR-5MS GC Columns
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt