missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ TraceGOLD TG-200MS GC Column
Thermo Scientific™ TraceGOLD™ TG-200MS GC Column has an exceptionally inert phase with selectivity and elution order optimized for difficult separations.
860.64 € - 1438.51 €
Product Code | Brand | Length (Metric) | Diameter (Metric) Inner | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) Inner | Price | Quantity & Availability | |||||
15820900
|
Thermo Scientific™
26084-2980 |
30 m | 0.53 mm |
860.64 €
Each |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||||
15547791
|
Thermo Scientific™
26084-3080 |
60 m | 0.25 mm |
1438.51 €
Each |
Estimated Shipment: 15-05-2025
Log in to see stock. |
|||||
Description
- Trifluoropropylmethyl polysiloxane phase resolves compounds that phenyl and cyano phases cannot
- Outstanding thermal stability and low bleed
- Suitable for use with sensitive detectors including ECD, NPD and MS
- Appropriate for use as a confirmation GC column
Specifications
G6 | |
TraceGOLD TG-200MS | |
1 μm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ TraceGOLD TG-200MS GC Column