Learn More
Thermo Scientific™ TraceGOLD TG-5SilMS GC Columns
Incorporates phenyl groups in the polymer backbone for improved thermal stability, reduced bleed and reduced susceptibility to oxidation.
335.00 € - 1320.88 €
Specifications
Stationary Phase | TraceGOLD™ TG-5 SilMS |
---|---|
USP Type | G27, G36 |
Max. Temperature | 330/350°C |
For Use With (Application) | GC/MS Applications Using Ion-Trap Systems, GC/MS for Polycyclic Aromatics, Phthalates, Phenols, Amines, and Organophosphates, GC/MS for Hydrocarbons Including Chlorinated Hydrocarbons |
Polarity | Low-Polar |
Product Code | Brand | Length (Metric) | Diameter (Metric) Inner | Film Thickness | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) Inner | Film Thickness | Price | Quantity & Availability | |||||
11521174
|
Thermo Scientific™
26096-5800 |
40 m | 0.18 mm | 0.18 μm | N/A | ||||||
13418018
|
Thermo Scientific™
26096-2760 |
20 m | 0.15 mm | 0.15 μm |
984.20 €
Each |
Estimated Shipment: 26-05-2025
Log in to see stock. |
|||||
15297478
|
Thermo Scientific™
26096-5820 |
12 m | 0.2 mm | 0.33 μm | N/A | ||||||
13498008
|
Thermo Scientific™
26096-2750 |
10 m | 0.15 mm | 0.15 μm | N/A | ||||||
10311595
|
Thermo Scientific™
26096-1300 |
15 m | 0.25 mm | 0.25 μm |
482.44 €
Each |
Estimated Shipment: 28-04-2025
Log in to see stock. |
|||||
10794637
|
Thermo Scientific™
26096-1310 |
15 m | 0.32 mm | 0.25 μm | N/A | ||||||
10177894
|
Thermo Scientific™
26096-1420 |
30 m | 0.25 mm | 0.25 μm |
793.95 €
Each |
In Stock
Log in to see stock. |
|||||
10457524
|
Thermo Scientific™
26096-1430 |
30 m | 0.32 mm | 0.25 μm |
822.28 €
Each |
In Stock
Log in to see stock. |
|||||
10526005
|
Thermo Scientific™
26096-1540 |
60 m | 0.25 mm | 0.25 μm |
1304.42 €
Each |
Estimated Shipment: 18-04-2025
Log in to see stock. |
|||||
10586375
|
Thermo Scientific™
26096-2110 |
15 m | 0.25 mm | 0.5 μm | N/A | ||||||
10239184
|
Thermo Scientific™
26096-2230 |
30 m | 0.25 mm | 0.5 μm |
787.31 €
Each |
In Stock
Log in to see stock. |
|||||
10698905
|
Thermo Scientific™
26096-2240 |
30 m | 0.32 mm | 0.5 μm |
838.40 €
Each |
Estimated Shipment: 24-04-2025
Log in to see stock. |
|||||
10260335
|
Thermo Scientific™
26096-2840 |
15 m | 0.25 mm | 1 μm | N/A | ||||||
10447914
|
Thermo Scientific™
26096-2960 |
30 m | 0.25 mm | 1 μm |
774.66 €
Each |
Estimated Shipment: 22-04-2025
Log in to see stock. |
|||||
10046034
|
Thermo Scientific™
26096-2970 |
30 m | 0.32 mm | 1 μm |
832.66 €
Each |
Estimated Shipment: 02-05-2025
Log in to see stock. |
|||||
10167664
|
Thermo Scientific™
26096-3080 |
60 m | 0.25 mm | 1 μm |
1320.88 €
Each |
Estimated Shipment: 24-04-2025
Log in to see stock. |
|||||
10702347
|
Thermo Scientific™
26096-3360 |
30 m | 0.53 mm | 1.5 μm |
962.17 €
Each |
Estimated Shipment: 24-04-2025
Log in to see stock. |
|||||
13178295
|
Thermo Scientific™
26096-5780 |
20 m | 0.18 mm | 0.18 μm |
706.61 €
Each |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||||
Description
Thermo Scientific™ TraceGOLD™ TG-5SilMS GC Columns incorporate phenyl groups in the polymer backbone for improved thermal stability, reduced bleed and reduced susceptibility to oxidation.
- Low polarity, silarylene phase with selectivity comparable to 5% diphenyl/95% dimethyl polysiloxane
- Designed for low bleed and outstanding inertness
- Similar to DB-5MS, VF-5ms, CP-Sil 8 Low-Bleed/MS, Rxi-5Sil MS, BPX5, ZB-5ms, Optima-5MS, SLB-5
Recommended for:
- GC/MS applications using ion-trap systems
- Polycyclic aromatics
- Hydrocarbons including chlorinated hydrocarbons
- Phthalates
- Phenols
- Amines
- Organophosphates
Specifications
TraceGOLD™ TG-5 SilMS | |
330/350°C | |
Low-Polar |
G27, G36 | |
GC/MS Applications Using Ion-Trap Systems, GC/MS for Polycyclic Aromatics, Phthalates, Phenols, Amines, and Organophosphates, GC/MS for Hydrocarbons Including Chlorinated Hydrocarbons |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Thermo Scientific™ TraceGOLD TG-5SilMS GC Columns