missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-2-Undecenal, 95%, remainder mainly cis-isomer, Thermo Scientific™
47.00 € - 136.00 €
Chemical Identifiers
CAS | 53448-07-0 |
---|---|
Molecular Formula | C11H20O |
Molecular Weight (g/mol) | 168.28 |
MDL Number | MFCD00014680 |
InChI Key | PANBRUWVURLWGY-MDZDMXLPSA-N |
Synonym | trans-2-undecenal, 2-undecenal, e-undec-2-enal, e-2-undecenal, 2-undecenal, 2e, 2-undecen-1-al, 2e-2-undecenal, trans-2-undecen-1-al, 2-undecenal, e, unii-fnp3s9mg30 |
PubChem CID | 5283356 |
IUPAC Name | (E)-undec-2-enal |
SMILES | CCCCCCCCC=CC=O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
53448-07-0 | |
168.28 | |
PANBRUWVURLWGY-MDZDMXLPSA-N | |
5283356 | |
CCCCCCCCC=CC=O |
C11H20O | |
MFCD00014680 | |
trans-2-undecenal, 2-undecenal, e-undec-2-enal, e-2-undecenal, 2-undecenal, 2e, 2-undecen-1-al, 2e-2-undecenal, trans-2-undecen-1-al, 2-undecenal, e, unii-fnp3s9mg30 | |
(E)-undec-2-enal |
Specifications
53448-07-0 | |
115°C (10 mmHg) | |
C11H20O | |
MFCD00014680 | |
Air sensitive | |
PANBRUWVURLWGY-MDZDMXLPSA-N | |
(E)-undec-2-enal | |
5283356 | |
95% | |
trans-2-Undecenal |
0.844 | |
104°C (219°F) | |
1.457 | |
1762239 | |
trans-2-undecenal, 2-undecenal, e-undec-2-enal, e-2-undecenal, 2-undecenal, 2e, 2-undecen-1-al, 2e-2-undecenal, trans-2-undecen-1-al, 2-undecenal, e, unii-fnp3s9mg30 | |
CCCCCCCCC=CC=O | |
168.28 | |
168.28 | |
remainder mainly cis-isomer |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
missing translation for 'einecsNumber' : 219-564-6
missing translation for 'tsca' : No
Recommended Storage : Keep cold