Learn More
trans-3-Fluorocinnamic acid, 98%, Thermo Scientific Chemicals
33.70 € - 428.00 €
Chemical Identifiers
CAS | 20595-30-6 |
---|---|
Molecular Formula | C9H7FO2 |
Molecular Weight (g/mol) | 166.151 |
MDL Number | MFCD00004383 |
InChI Key | RTSIUKMGSDOSTI-SNAWJCMRSA-N |
Synonym | 3-fluorocinnamic acid, 3-3-fluorophenyl acrylic acid, trans-3-fluorocinnamic acid, 2e-3-3-fluorophenyl prop-2-enoic acid, e-3-3-fluorophenyl acrylic acid, m-fluorocinnamic acid, trans 3-fluorocinnamic acid, 3-3-fluoro-phenyl-acrylic acid, trans-m-fluorocinnamic, e-3-3-fluorophenyl prop-2-enoic acid |
PubChem CID | 1551219 |
IUPAC Name | (E)-3-(3-fluorophenyl)prop-2-enoic acid |
SMILES | C1=CC(=CC(=C1)F)C=CC(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11465746
|
Thermo Scientific Alfa Aesar
B20818.06 |
5 g |
33.70 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11475746
|
Thermo Scientific Alfa Aesar
B20818.14 |
25 g |
116.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11485746
|
Thermo Scientific Alfa Aesar
B20818.22 |
100 g |
428.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
20595-30-6 | |
166.151 | |
RTSIUKMGSDOSTI-SNAWJCMRSA-N | |
1551219 | |
C1=CC(=CC(=C1)F)C=CC(=O)O |
C9H7FO2 | |
MFCD00004383 | |
3-fluorocinnamic acid, 3-3-fluorophenyl acrylic acid, trans-3-fluorocinnamic acid, 2e-3-3-fluorophenyl prop-2-enoic acid, e-3-3-fluorophenyl acrylic acid, m-fluorocinnamic acid, trans 3-fluorocinnamic acid, 3-3-fluoro-phenyl-acrylic acid, trans-m-fluorocinnamic, e-3-3-fluorophenyl prop-2-enoic acid | |
(E)-3-(3-fluorophenyl)prop-2-enoic acid |
Specifications
20595-30-6 | |
Odorless | |
MFCD00004383 | |
3-fluorocinnamic acid, 3-3-fluorophenyl acrylic acid, trans-3-fluorocinnamic acid, 2e-3-3-fluorophenyl prop-2-enoic acid, e-3-3-fluorophenyl acrylic acid, m-fluorocinnamic acid, trans 3-fluorocinnamic acid, 3-3-fluoro-phenyl-acrylic acid, trans-m-fluorocinnamic, e-3-3-fluorophenyl prop-2-enoic acid | |
C1=CC(=CC(=C1)F)C=CC(=O)O | |
166.151 | |
166.15 | |
trans-3-Fluorocinnamic acid |
165°C to 168°C | |
C9H7FO2 | |
1365172 | |
RTSIUKMGSDOSTI-SNAWJCMRSA-N | |
(E)-3-(3-fluorophenyl)prop-2-enoic acid | |
1551219 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 207-281-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only