missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-Zeatin (synthetic), 97+%, Thermo Scientific Chemicals
103.52 € - 606.02 €
Chemical Identifiers
CAS | 1637-39-4 |
---|---|
Molecular Formula | C10H13N5O |
Molecular Weight (g/mol) | 219.25 |
MDL Number | MFCD00213654 |
InChI Key | UZKQTCBAMSWPJD-FARCUNLSSA-N |
Synonym | trans-zeatin, zeatin, e-zeatin, zeatine, e-2-methyl-4-1h-purin-6-ylamino-2-buten-1-ol, trans-zeatin synthetic, unii-7i6ooj9gr6, 2-buten-1-ol, 2-methyl-4-1h-purin-6-ylamino-, 2e, 2e-2-methyl-4-9h-purin-6-ylamino but-2-en-1-ol, 2-buten-1-ol, 2-methyl-4-1h-purin-6-ylamino-, e |
PubChem CID | 449093 |
ChEBI | CHEBI:16522 |
IUPAC Name | (E)-2-methyl-4-(7H-purin-6-ylamino)but-2-en-1-ol |
SMILES | CC(=CCNC1=NC=NC2=C1NC=N2)CO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10697231
|
Thermo Scientific Acros
264290100 |
10 mg | Glass bottle |
103.52 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10480711
|
Thermo Scientific Acros
264291000 |
100 mg | Glass bottle |
606.02 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1637-39-4 | |
219.25 | |
UZKQTCBAMSWPJD-FARCUNLSSA-N | |
449093 | |
(E)-2-methyl-4-(7H-purin-6-ylamino)but-2-en-1-ol |
C10H13N5O | |
MFCD00213654 | |
trans-zeatin, zeatin, e-zeatin, zeatine, e-2-methyl-4-1h-purin-6-ylamino-2-buten-1-ol, trans-zeatin synthetic, unii-7i6ooj9gr6, 2-buten-1-ol, 2-methyl-4-1h-purin-6-ylamino-, 2e, 2e-2-methyl-4-9h-purin-6-ylamino but-2-en-1-ol, 2-buten-1-ol, 2-methyl-4-1h-purin-6-ylamino-, e | |
CHEBI:16522 | |
CC(=CCNC1=NC=NC2=C1NC=N2)CO |
Specifications
1637-39-4 | |
Colorless to Yellow | |
Authentic | |
C10H13N5O | |
15, 10315 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol and dimethylformamide | |
UZKQTCBAMSWPJD-FARCUNLSSA-N | |
(E)-2-methyl-4-(7H-purin-6-ylamino)but-2-en-1-ol | |
449093 | |
219.25 | |
2% max. (HPLC) | |
trans-Zeatin (synthetic), 97% |
207°C | |
1% max. | |
97% min. (HPLC) | |
MFCD00213654 | |
trans-zeatin, zeatin, e-zeatin, zeatine, e-2-methyl-4-1h-purin-6-ylamino-2-buten-1-ol, trans-zeatin synthetic, unii-7i6ooj9gr6, 2-buten-1-ol, 2-methyl-4-1h-purin-6-ylamino-, 2e, 2e-2-methyl-4-9h-purin-6-ylamino but-2-en-1-ol, 2-buten-1-ol, 2-methyl-4-1h-purin-6-ylamino-, e | |
96.0% min. (on dried substance) | |
CC(=CCNC1=NC=NC2=C1NC=N2)CO | |
219.25 | |
CHEBI:16522 | |
97+% | |
Liquid |
Safety and Handling
missing translation for 'rtecsNumber' : EM9506000
Product Identifier
- trans-Zeatin (synthetic)
RUO – Research Use Only