missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brand™ Multi-Channel Micropipette Transferpette™ -12 Electronic, DE-M
Ergonomic, handy design. Clear and intuitive operation. Up to 4,000 pipetting cycles on a single charge.
1280.00 €
Specifications
Autoclavable | Autoclavable |
---|---|
Number of Channels | 12 |
Product Line | Transferpette |
Programmability | Yes |
Product Code | Brand | Volume (Metric) | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Includes | Price | Quantity & Availability | ||||
12378623
|
Brand™
705466 |
15 to 300 μL | UK/Ireland Adapter |
1280.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12358623
|
Brand™
705463 |
5 to 100 μL | UK/Ireland AC Adapter |
1280.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11673229
|
Brand™
705464 |
10 to 200 μL | UK/Ireland AC Adapter |
1280.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10293212
|
Brand™
705456 |
15 to 300 μL | Europe AC Adapter |
1280.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10131932
|
Brand™
705453 |
5 to 100 μL | Europe AC Adapter |
1280.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11663229
|
Brand™
705459 |
1 to 20 μL | EU plug |
1280.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12634060
|
Brand™
705460 |
1 to 20 μL | UK/Ireland Adapter |
1280.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Transferpette electronic pipettes combine the globally proven properties of mechanical piston operated air displacement pipettes by BRAND with the advantages of an electronic device.
- Features: The pipette shaft can be unscrewed and is entirely autoclavable (121 °C).
- Functions: pipetting, reverse pipetting, mixing, dispensing, up to 300 μl also GEL-Electrophoresis.
Easy Calibration-Technique for making quick adjustments of the Transferpette™ electronic, without tools. - Limitations of use: Operating temperature of instrument and reagent should be between +15 °C and +40 °C (59 °F to 104 °F) (other temperatures upon request)
- Vapor pressure up to 500 mbar
- Viscosity: 260 mPa s
- For viscous media, the speed must be adjusted if necessary.
Specifications
Autoclavable | |
12 | |
Transferpette | |
Yes |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Brand™ Multi-Channel Micropipette Transferpette™ -12 Electronic, DE-M