missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Multi-Channel Micropipette Transferpette™ -8 Electronic, DE-M

Ergonomic, handy design. Clear and intuitive operation. Up to 4,000 pipetting cycles on a single charge.

Brand:  BRAND™ 705414

844.35 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 11613229

  • 1105.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Transferpette electronic pipettes combine the globally proven properties of mechanical piston operated air displacement pipettes by BRAND with the advantages of an electronic device.
  • Features: The pipette shaft can be unscrewed and is entirely autoclavable (121 °C).
  • Functions: pipetting, reverse pipetting, mixing, dispensing, up to 300 μl also GEL-Electrophoresis.
    Easy Calibration-Technique for making quick adjustments of the Transferpette™ electronic, without tools.
  • Limitations of use: Operating temperature of instrument and reagent should be between +15 °C and +40 °C (59 °F to 104 °F) (other temperatures upon request)
  • Vapor pressure up to 500 mbar
  • Viscosity: 260 mPa s
  • For viscous media, the speed must be adjusted if necessary.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

≤ 0.8%, ± 1.6μL
Autoclavable
8
Transferpette
Multi-Channel Pipette
LCD
UK/Ireland AC Adapter
1 Each
230 V
360°
Yellow
8
Yes
10 to 200 μL
Ergonomic
0.2μL
230V/50 Hz
50 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BRAND™ Multi-Channel Micropipette Transferpette™ -8 Electronic, DE-M > Volume: 10 to 200uL, Model 8, ac adapter for UK/Ireland

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.