missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Multi-Channel Micropipette Transferpette™ -8 Electronic, DE-M
Ergonomic, handy design. Clear and intuitive operation. Up to 4,000 pipetting cycles on a single charge.
1090.00 € - 1105.00 €
Specifications
Angle | 360° |
---|---|
Autoclavable | Autoclavable |
Model | 8 |
Number of Channels | 8 |
Product Line | Transferpette |
Product Code | Brand | Volume (Metric) | Color | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Color | Includes | Price | Quantity & Availability | |||
12398603
|
BRAND™
705410 |
1 to 20 μL | Gray | UK/Ireland AC Adapter |
1105.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10797444
|
BRAND™
705404 |
10 to 200 μL | Yellow | Europe AC Adapter |
1090.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12328613
|
BRAND™
705416 |
15 to 300 μL | Green | UK/Ireland AC Adapter |
1105.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12308613
|
BRAND™
705413 |
5 to 100 μL | Yellow | UK/Ireland AC Adapter |
1105.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11613229
|
BRAND™
705414 |
10 to 200 μL | Yellow | UK/Ireland AC Adapter |
1105.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11603229
|
BRAND™
705409 |
0.5 to 10 μL | Gray | UK/Ireland AC Adapter |
1105.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10233172
|
BRAND™
705406 |
15 to 300 μL | Green | Europe AC Adapter |
1105.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10477852
|
BRAND™
705400 |
1 to 20 μL | Gray | Europe AC Adapter |
1105.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10191972
|
BRAND™
705403 |
5 to 100 μL | Yellow | Europe AC Adapter |
1105.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Transferpette electronic pipettes combine the globally proven properties of mechanical piston operated air displacement pipettes by BRAND with the advantages of an electronic device.
- Features: The pipette shaft can be unscrewed and is entirely autoclavable (121 °C).
- Functions: pipetting, reverse pipetting, mixing, dispensing, up to 300 μl also GEL-Electrophoresis.
Easy Calibration-Technique for making quick adjustments of the Transferpette™ electronic, without tools. - Limitations of use: Operating temperature of instrument and reagent should be between +15 °C and +40 °C (59 °F to 104 °F) (other temperatures upon request)
- Vapor pressure up to 500 mbar
- Viscosity: 260 mPa s
- For viscous media, the speed must be adjusted if necessary.
Specifications
360° | |
8 | |
Transferpette | |
LCD | |
1 Each |
Autoclavable | |
8 | |
Yes | |
Ergonomic | |
230V/50 Hz |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BRAND™ Multi-Channel Micropipette Transferpette™ -8 Electronic, DE-M