Learn More
Fisherbrand™ Transparent Terylene Mesh PVC Tubing
General purpose tubing for transfer of fluids, gases, and powdered foods
Brand:  Fisherbrand™ RPVCNT/9.5/10MT
 View more versions of this product
Product Code. 12424080
  • 30.00 € / 10m
Estimated Shipment: 13-12-2024
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
All dimensions are approximate.
  • Transparent, colorless
  • Terylene mesh reinforced wall
  • Flexible yet great inherent strength
  • Suitable for moderate pressure or vacuum applications
Specifications
Specifications
10 x 3mm
10 m
1 Pack
PVC
Suitable For Moderate Pressure Or Vacuum Applications
Transparent