missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tri(1-aziridinyl)phosphine sulfide, 98%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Solubility
Soluble in benzene, acetone and methanol.
Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
Soluble in benzene, acetone and methanol.
Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.

Specifications
Specifications
Quantity | 5 g |
UN Number | UN2811 |
Solubility Information | Soluble in benzene,acetone and methanol. |
Formula Weight | 189.22 |
Percent Purity | 98% |
Physical Form | Solid |
Chemical Name or Material | Tri(1-aziridinyl)phosphine sulfide |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 7
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tri(1-aziridinyl)phosphine sulfide, 98%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction