missing translation for 'onlineSavingsMsg'
Learn More
Learn More
β-Tri-Calcium phosphate, ≥98% β to phase basis, Honeywell Fluka™
Puriss. p.a.,≥98% β-phase basis (sintered Powder)
146.44 € - 765.20 €
Chemical Identifiers
CAS | 7758-87-4 |
---|---|
Molecular Formula | Ca3O8P2 |
Molecular Weight (g/mol) | 310.174 |
MDL Number | MFCD00015984 |
InChI Key | QORWJWZARLRLPR-UHFFFAOYSA-H |
Synonym | calcium phosphate, synthos, tricalcium phosphate, tricalcium diphosphate, calcigenol simple, calcium orthophosphate, beta-tcp, tribasic calcium phosphate, bonarka, calcium phosphate, tribasic |
PubChem CID | 24456 |
ChEBI | CHEBI:9679 |
IUPAC Name | tricalcium;diphosphate |
SMILES | [O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Ca+2].[Ca+2].[Ca+2] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7758-87-4 | |
310.174 | |
QORWJWZARLRLPR-UHFFFAOYSA-H | |
24456 | |
tricalcium;diphosphate |
Ca3O8P2 | |
MFCD00015984 | |
calcium phosphate, synthos, tricalcium phosphate, tricalcium diphosphate, calcigenol simple, calcium orthophosphate, beta-tcp, tribasic calcium phosphate, bonarka, calcium phosphate, tribasic | |
CHEBI:9679 | |
[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Ca+2].[Ca+2].[Ca+2] |
Specifications
1670°C | |
White | |
Ca3O8P2 | |
NONH for all modes of transport | |
QORWJWZARLRLPR-UHFFFAOYSA-H | |
tricalcium;diphosphate | |
24456 | |
310.18g/mol | |
3.27g/cm3 at 20.5°C |
7758-87-4 | |
Solid | |
MFCD00015984 | |
calcium phosphate, synthos, tricalcium phosphate, tricalcium diphosphate, calcigenol simple, calcium orthophosphate, beta-tcp, tribasic calcium phosphate, bonarka, calcium phosphate, tribasic | |
[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Ca+2].[Ca+2].[Ca+2] | |
310.174 | |
CHEBI:9679 | |
≥98% β to phase basis (sintered Powder) | |
β-tri-Calcium phosphate |
Safety and Handling
missing translation for 'rtecsNumber' : 231-840-8
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title