missing translation for 'onlineSavingsMsg'
Learn More
Tri-n-octylphosphine oxide, 99%, Thermo Scientific Chemicals Product Code.: 10115453

Tri-n-octylphosphine oxide, 99%, Thermo Scientific Chemicals

Product Code. 10115453
500 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10115453

Brand: Thermo Scientific Acros 207605000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-50-2
Molecular Formula C24H51OP
Molecular Weight (g/mol) 386.64
MDL Number MFCD00002083
InChI Key ZMBHCYHQLYEYDV-UHFFFAOYSA-N
Synonym trioctylphosphine oxide, tri-n-octylphosphine oxide, topo, phosphine oxide, trioctyl, cyanex 921, hostarex px 324, trioctyl phosphine oxide, trioctylphosphane oxide, trioctylphosphino-1-one, acmc-209peg
PubChem CID 65577
IUPAC Name 1-dioctylphosphoryloctane
SMILES CCCCCCCCP(=O)(CCCCCCCC)CCCCCCCC

Specifications

Melting Point 50.0°C to 55.0°C
Color White to Yellow
Boiling Point 201.0°C to 202.0°C (2.0 mmHg)
Flash Point 230°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (including isomer) (GC)
Packaging Glass bottle
Linear Formula [CH3(CH2)7]3P(O)
Quantity 500 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in cyclohexane,methanol
Formula Weight 386.64
Percent Purity 99%
Physical Form Crystalline Powder, Crystals or Flakes
Chemical Name or Material Tri-n-octylphosphine oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467319 Certificate TypeCertificate of Analysis Date01/11/2024 Catalog Number207605000,207600250,207601000
Lot NumberA0466187 Certificate TypeCertificate of Analysis Date16/09/2024 Catalog Number207605000,207600250,207601000
Lot NumberA0463093 Certificate TypeCertificate of Analysis Date27/05/2024 Catalog Number207605000,207600250,207601000
Lot NumberA0461145 Certificate TypeCertificate of Analysis Date15/03/2024 Catalog Number207605000,207600250,207601000
Lot NumberA0444532 Certificate TypeCertificate of Analysis Date29/01/2024 Catalog Number207605000,207600250,207601000
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tri-n-octylphosphine oxide, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.