Learn More
Thermo Scientific™ Tri-Sil TBT (TMSI:BSA:TMCS) Reagent
Employ a catalyzed silylation reagent formulation, for derivatization of hydroxyls with the Thermo Scientific™ Tri-Sil TBT (TMSI:BSA:TMCS) Reagent.
Brand:  Thermo Scientific™ TS-49016
411.02 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11537851
  • 413.08 € / Pack of 10
Explore more special offers
Description
Description
Employ a catalyzed silylation reagent formulation, for derivatization of hydroxyls with the Thermo Scientific™ Tri-Sil TBT (TMSI:BSA:TMCS) Reagent. Composed of TMSI/BSA/TMCS (3:3:2), this catalyzed silylation reagent formulation typically reacts with hydroxyls in a short period of time at 60–80 °C to form TMS ethers. Highly hindered hydroxyls may require several hours.
  • Converts all classes of hydroxyl groups to TMS ethers
  • Under usual conditions, the reaction is complete in a short period of time at 60 to 80°C, although very hindered hydroxyls may require several hours
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GC
25 - 45, 25 - 45, 20 - 25%
10 x 1 mL ampules
Toxic
Room Temperature
Trimethylsilyl N-trimethylsilylacetamidate, 1-(Trimethylsilyl)-1 H-imidazole, Chlorotrimethylsilane
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Tri-Sil TBT (TMSI:BSA:TMCS) Reagent > 10 x 1mL ampules