missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Triangular Tripod Stand

Designed to cope with the demands of a modern laboratory.

Brand:  Fisherbrand™ 3001

 View more versions of this product

Product Code. 11779806

  • 13.80 € / Each
Estimated Shipment: 24-03-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

Zinc plated steel with 210mm height, 6mm diameter legs.
Specifications

Specifications

Stainless Steel
125 mm side length
Triangular
210 mm
Tripod Stand
Zinc Plated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.