Learn More
Tributylamine, 99%, Thermo Scientific Chemicals
28.84 € - 627.00 €
Chemical Identifiers
CAS | 102-82-9 |
---|---|
Molecular Formula | C12H27N |
Molecular Weight (g/mol) | 185.35 |
MDL Number | MFCD00009431 |
InChI Key | IMFACGCPASFAPR-UHFFFAOYSA-N |
Synonym | tributylamine, tri-n-butylamine, 1-butanamine, n,n-dibutyl, n,n-dibutyl-1-butanamine, tributilamina, tris-n-butylamine, amine, tributyl, tris n-butylamine, tributilamina romanian, unii-c3tzb2w0r7 |
PubChem CID | 7622 |
ChEBI | CHEBI:38905 |
IUPAC Name | N,N-dibutylbutan-1-amine |
SMILES | CCCCN(CCCC)CCCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10447340
|
Thermo Scientific Acros
139321000 |
100 mL | Glass bottle |
28.84 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10387053
|
Thermo Scientific Acros
139322500 |
250 mL | Glass bottle |
46.43 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10133190
|
Thermo Scientific Acros
139320010 |
1 L | Glass bottle |
102.51 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10377053
|
Thermo Scientific Acros
139320025 |
2.5 L | Glass bottle |
178.89 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11975171
|
Thermo Scientific Acros
139320100 |
10 L | Plastic drum |
627.00 €
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
102-82-9 | |
185.35 | |
IMFACGCPASFAPR-UHFFFAOYSA-N | |
7622 | |
N,N-dibutylbutan-1-amine |
C12H27N | |
MFCD00009431 | |
tributylamine, tri-n-butylamine, 1-butanamine, n,n-dibutyl, n,n-dibutyl-1-butanamine, tributilamina, tris-n-butylamine, amine, tributyl, tris n-butylamine, tributilamina romanian, unii-c3tzb2w0r7 | |
CHEBI:38905 | |
CCCCN(CCCC)CCCC |
Specifications
102-82-9 | |
0.7780g/mL | |
75°C | |
98.5% min. (GC) | |
[CH3NCH2)3]3N | |
MFCD00009431 | |
01,1189; 06,601 | |
0.778 | |
Solubility in water: 0.08g/L water (20°C). Other solubilities: soluble in most organic solvents,very soluble in alcohol and ether | |
CCCCN(CCCC)CCCC | |
185.35 | |
CHEBI:38905 | |
185.35 | |
Liquid |
-70.0°C | |
215.0°C | |
Authentic | |
C12H27N | |
1.4273 to 1.4293 | |
04,157 | |
15,978 | |
tributylamine, tri-n-butylamine, 1-butanamine, n,n-dibutyl, n,n-dibutyl-1-butanamine, tributilamina, tris-n-butylamine, amine, tributyl, tris n-butylamine, tributilamina romanian, unii-c3tzb2w0r7 | |
IMFACGCPASFAPR-UHFFFAOYSA-N | |
N,N-dibutylbutan-1-amine | |
7622 | |
2.4 mPa.s (20°C) | |
99% | |
Tributylamine |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Fatal in contact with skin.
Causes skin irritation.
Fatal if inhaled.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position c
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-058-7
- Tributylamine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H315-Causes skin irritation.
Precautionary Statement
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P310-Immediately call a POISON CENTER/doctor/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
RUO – Research Use Only