missing translation for 'onlineSavingsMsg'
Learn More
Trifluoroacetic acid, 99+%, extra pure, Thermo Scientific Chemicals Product Code.: 10031550

Trifluoroacetic acid, 99+%, extra pure, Thermo Scientific Chemicals

Product Code. 10031550
25 g, Glass Bottle
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
2.5 kg
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 10031550

Brand: Thermo Scientific Acros 139720250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-05-1
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
Synonym trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctovaShow MoreShow Less
PubChem CID 6422
ChEBI CHEBI:45892
SMILES OC(=O)C(F)(F)F

Specifications

Melting Point -15.0°C
Boiling Point 72.0°C
Color Colorless
Assay Percent Range 0.99
Linear Formula CF3CO2H
Beilstein 02, II, 186
Fieser 01,1219; 02,433; 03,305; 04,530; 05,695; 06,613; 07,388; 08,503; 09,483; 10,418; 11,557; 12,529; 14,322; 15,338; 16,83; 17,340
Merck Index 15, 9847
Solubility Information Solubility in water: miscible. Other solubilities: miscible with ether, acetone, ethanol, benzene,, ccl4, hexane, methanol, xylene and hexane
IUPAC Name 2,2,2-trifluoroacetic acid
Formula Weight 114.02
Percent Purity 99%
Grade Extra Pure
Density 1.5350 g/mL
Quantity 25 g
Infrared Spectrum Conforms to Reference
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3
Vapor Pressure 107 mbar at 25°C
Chemical Name or Material Trifluoroacetic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0458514 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number139725000,139720250,139720100,139720010,139721000,139720025
Lot NumberA0465092 Certificate TypeCertificate of Analysis Date01/08/2024 Catalog Number139725000,139720250,139720100,139720010,139721000,139720025
Lot NumberA0462766 Certificate TypeCertificate of Analysis Date13/05/2024 Catalog Number139725000,139720250,139720100,139720010,139721000,139720025
Lot NumberA0448760 Certificate TypeCertificate of Analysis Date20/12/2023 Catalog Number139725000,139720250,139720100,139720010,139721000,139720025
Lot NumberA0458514 Certificate TypeCertificate of Analysis Date13/12/2023 Catalog Number139725000,139720250,139720100,139720010,139721000,139720025
compliance-icons compliance-icons
Product Identifier
  • Trifluoroacetic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • H332-Harmful if inhaled.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Trifluoroacetic acid, 99+%, extra pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.