missing translation for 'onlineSavingsMsg'
Learn More

Trimethylamine, 45% w/w aq. soln., Thermo Scientific Chemicals

95.78 € - 277.38 €

Chemical Identifiers

CAS 75-50-3
Molecular Formula C3H9N
Molecular Weight (g/mol) 59.11
MDL Number MFCD00008327
InChI Key GETQZCLCWQTVFV-UHFFFAOYSA-N
Synonym trimethylamine, methanamine, n,n-dimethyl, dimethylmethaneamine, n-trimethylamine, trimethylamine solution, ch3 3n, trimethyl amine, trimethylamin, trimethyl-amine, fema number 3241
PubChem CID 1146
ChEBI CHEBI:18139
SMILES CN(C)C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11336219
View Documents
Thermo Scientific Alfa Aesar
043282.AP
500 mL
95.78 €
500mL
Please to purchase this item. Need a web account? Register with us today!
11319408
View Documents
Thermo Scientific Alfa Aesar
043282.K4
2.5 L
277.38 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Trimethylamine solution used in research. As flavoring agents and adjuvants. Trimethylamine is used in the synthesis of choline, tetramethylammonium hydroxide, plant growth regulators, strongly basic anion exchange resins, dye leveling agents and a number of basic dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

75-50-3
59.11
GETQZCLCWQTVFV-UHFFFAOYSA-N
1146
CN(C)C
C3H9N
MFCD00008327
trimethylamine, methanamine, n,n-dimethyl, dimethylmethaneamine, n-trimethylamine, trimethylamine solution, ch3 3n, trimethyl amine, trimethylamin, trimethyl-amine, fema number 3241
CHEBI:18139

Specifications

75-50-3
Colorless
−20°C (−4°F)
C3H9N
UN1297
14,9710
Soluble In Water,8.9e+005mg/L.
CN(C)C
59.11
CHEBI:18139
45% w/w aq. soln.
Trimethylamine
-2°C
30°C
Unpleasant
MFCD00008327
956566
trimethylamine, methanamine, n,n-dimethyl, dimethylmethaneamine, n-trimethylamine, trimethylamine solution, ch3 3n, trimethyl amine, trimethylamin, trimethyl-amine, fema number 3241
GETQZCLCWQTVFV-UHFFFAOYSA-N
trimethylamine
1146
59.11
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H224-H314-H302-H332-H335
Extremely flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H224-H302+H332-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: TRIMETHYLAMINE, AQUEOUS SOLUTIONS

missing translation for 'einecsNumber' : 200-875-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only