Learn More
Thermo Scientific™ Triple Wrap Sterile Pack w/VHP Indicator Tryptone Soya Agar
Carry out environmental monitoring in your most regulated environments using Thermo Scientific™ Triple Wrapped Plates, for passive and active air sampling applications.
Brand: Thermo Scientific™ PO5510D
Description
Experience next-level cleanroom confidence with Thermo Scientific™ Triple Wrap Sterile Pack Tryptone Soya Agar plates, complete with VHP Indicator.
Achieve confidence in your results and ensure optimum media fertility using plates with a Vapour-phase Hydrogen Peroxide (VHP) exposure indicator, designed to confirm the integrity of the wrapping material and seal during VHP exposure.Three protective layers reduce risk of contamination:
- Primary film with moisture control patch
- Secondary film VHP barrier with immobilized desiccant sachet
- Tertiary film dust cover
- The incorporation of novel elements enhance moisture control by permitting the passage and absorption of excess moisture
- Using plates with an improved shelf life and room temperature storage offers a cost-effective solution, minimizing new batch validation and refrigeration costs
Not all products are available for sale in all territories. Please inquire.

Specifications
Ivory, transparent | |
55mm Contact Plate | |
Tryptone Soya Agar | |
13.0g ± 0.5g | |
100/Pk. |
2 to 25°C | |
Germany | |
Agar | |
Triple Wrapped - Irradiated |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6246025 | Certificate of Analysis | 24/03/2025 | PO5510D |
6228549 | Certificate of Analysis | 06/02/2025 | PO5510D |
6207594 | Certificate of Analysis | 12/12/2024 | PO5510D |
6198339 | Certificate of Analysis | 20/11/2024 | PO5510D |
6180907 | Certificate of Analysis | 19/10/2024 | PO5510D |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.