Learn More
Thermo Scientific™ TriPlus RSH Autosampler Fixed Needle Syringe, 10μL
Thermo Scientific™ TriPlus RSH Autosampler Fixed Needle Syringes are accurate and reproducible over a range of volumes.
Brand: Thermo Scientific™ 365D0271
78.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Thermo Scientific™ TriPlus RSH Autosampler Fixed Needle Syringes are accurate and reproducible over a range of volumes.
- Borosilicate glass with metal flanges
- Needles and plungers are stainless steel
- Field-proven syringe design
Specifications
Merlin Valve Injector, PTV and Split | |
GC Autosampler Syringe, Fixed Needle | |
57 mm | |
TriPlus RSH Autosampler and TriPlus 100LS Autosampler |
23s G | |
10 μL | |
Cone |