missing translation for 'onlineSavingsMsg'
Learn More
Tris(2,2'-bipyridyl)ruthenium(II) chloride, hexahydrate, 98%, Thermo Scientific Chemicals Product Code.: 10154740

Tris(2,2'-bipyridyl)ruthenium(II) chloride, hexahydrate, 98%, Thermo Scientific Chemicals

Product Code. 10154740
1 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10154740

Brand: Thermo Scientific Acros 208750010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50525-27-4
Molecular Formula C30H36Cl2N6O6Ru
Molecular Weight (g/mol) 748.62
MDL Number MFCD00149670
InChI Key WHELTKFSBJNBMQ-UHFFFAOYSA-L
Synonym tris 2,2'-bipyridyl ruthenium ii chloride hexahydrate
PubChem CID 131664161
IUPAC Name 2-pyridin-2-ylpyridine;ruthenium(2+);dichloride;hydrate
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Ru++].C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1

Specifications

Melting Point >300.0°C
Elemental Analysis 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: soluble.
Formula Weight 748.63
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Tris(2, 2'-bipyridyl)ruthenium(II) chloride, hexahydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465480 Certificate TypeCertificate of Analysis Date28/08/2024 Catalog Number208750010,208750050
Lot NumberA0456411 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog Number208750010,208750050
Lot NumberA0448776 Certificate TypeCertificate of Analysis Date03/02/2023 Catalog Number208750010,208750050
Lot NumberA0447397 Certificate TypeCertificate of Analysis Date03/02/2023 Catalog Number208750010,208750050
Lot NumberA0392753 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number208750010,208750050

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tris(2,2'-bipyridyl)ruthenium(II) chloride, hexahydrate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.