missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,2'-bipyridyl)ruthenium(II) chloride, hexahydrate, 98%, Thermo Scientific Chemicals

136.00 € - 464.00 €

Chemical Identifiers

CAS 50525-27-4
Molecular Formula C30H36Cl2N6O6Ru
Molecular Weight (g/mol) 748.62
MDL Number MFCD00149670
InChI Key WHELTKFSBJNBMQ-UHFFFAOYSA-L
Synonym tris 2,2'-bipyridyl ruthenium ii chloride hexahydrate
PubChem CID 131664161
IUPAC Name 2-pyridin-2-ylpyridine;ruthenium(2+);dichloride;hydrate
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Ru++].C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10154740
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Acros
208750010
1 g Glass bottle
136.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
10229203
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Acros
208750050
5 g Glass bottle
464.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

50525-27-4
748.62
WHELTKFSBJNBMQ-UHFFFAOYSA-L
131664161
O.O.O.O.O.O.[Cl-].[Cl-].[Ru++].C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1
C30H36Cl2N6O6Ru
MFCD00149670
tris 2,2'-bipyridyl ruthenium ii chloride hexahydrate
2-pyridin-2-ylpyridine;ruthenium(2+);dichloride;hydrate

Specifications

50525-27-4
97.5% min.
98%
MFCD00149670
Solubility in water: soluble.
O.O.O.O.O.O.[Cl-].[Cl-].[Ru++].C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1
748.62
748.63
Crystalline Powder or Crystals
>300.0°C
Authentic
C30H36Cl2N6O6Ru
tris 2,2'-bipyridyl ruthenium ii chloride hexahydrate
WHELTKFSBJNBMQ-UHFFFAOYSA-L
2-pyridin-2-ylpyridine;ruthenium(2+);dichloride;hydrate
131664161
98%
Tris(2, 2'-bipyridyl)ruthenium(II) chloride, hexahydrate
Videos
SDS
Documents

Documents

Certificates

RUO – Research Use Only