missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBE Buffer, Tris-Borate-EDTA, 1X Solution, Electrophoresis, Fisher BioReagents
Brand: Fisher BioReagents BP2430-20
194.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Save time and simplify your buffer preparation step by using ready-made Fisher BioReagents 1X TBE buffer solution
- Eliminates the hassle of dilution or waiting for powder to dissolve
- 1X solution contains 0.089M tris, 0.089M boric acid, and 0.002M EDTA
- Tested for the absence of DNase, RNase, and protease
- Filtered, autoclaved, and functionally tested for use in electrophoresis applications
- TBE is a commonly used buffer for electrophoresis of nucleic acids in agarose and polyacrylamide gels
- Intended for laboratory research use only
Specifications
Tris-Borate-EDTA | |
77-86-1, 6381-92-6, 10043-35-3, 7732-18-5 | |
8.2 to 8.4 | |
20 L | |
Filtered through a 0.2-micron filter. | |
Protease free | |
Electrophoresis |
1X Solution | |
Colorless | |
Liquid | |
DNase free | |
Poly CUBE | |
TBE |
Safety and Handling
Product Identifier
- Tris-borate-EDTA, 1X solution
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction