missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBE Buffer, Tris-Borate-EDTA, 1X Solution, Electrophoresis, Fisher BioReagents
42.50 € - 200.00 €
Specifications
Chemical Name or Material | Tris-Borate-EDTA |
---|---|
Name Note | 1X Solution |
CAS | 77-86-1, 6381-92-6, 10043-35-3, 7732-18-5 |
Color | Colorless |
pH | 8.2 to 8.4 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10754914
|
Fisher BioReagents
BP2430-1 |
1 L | Nalgene Poly Bottle |
42.50 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10755104
|
Fisher BioReagents
BP2430-20 |
20 L | Poly CUBE |
200.00 €
20 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Tris–Borate–EDTA (TBE) is commonly used as a buffer for nucleic acid electrophoresis.Specifications
Tris-Borate-EDTA | |
77-86-1, 6381-92-6, 10043-35-3, 7732-18-5 | |
8.2 to 8.4 | |
DNase free | |
Protease free | |
Electrophoresis |
1X Solution | |
Colorless | |
Liquid | |
Filtered through a 0.2-micron filter. | |
TBE |
Safety and Handling
Recommended Storage : RT
Product Identifier
- Tris-borate-EDTA, 1X solution
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title