missing translation for 'onlineSavingsMsg'
Learn More

Tris Buffer, 2M Solution (Molecular Biology), Fisher BioReagents

18.83 € - 41.41 €

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10376933
View Documents
Fisher BioReagents
BP1759-100
100 mL Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10705114
View Documents
Fisher BioReagents
BP1759-500
500 mL Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Tris is a buffer component in molecular biology, tissue culture, and electrophoresis procedures. Tris Solution, 2M, provides a convenient stock solution for preparing Tris buffers.
Specifications

Chemical Identifiers

77-86-1
121.136
6503
2-amino-2-(hydroxymethyl)propane-1,3-diol
C4H11NO3
LENZDBCJOHFCAS-UHFFFAOYSA-N
CHEBI:9754
C(C(CO)(CO)N)O

Specifications

77-86-1
1.95 to 2.05 M
DNase free
C4H11NO3
15, 9951
C(C(CO)(CO)N)O
121.136
CHEBI:9754
Tris Buffer, 2M Solution
Undesignated
Liquid
Pass Test
Protease free
LENZDBCJOHFCAS-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol
6503
Molecular Biology
Videos
Safety and Handling

Safety and Handling

Emergency Overview
May cause skin, eye, and respiratory tract irritation.
NFPA
Health: 1
Flammability:

Signal Word: CAUTION!

Recommended Storage : RT

Product Identifier
  • Tris buffer solution 2M

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.