missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris, MP Biomedicals™
Useful as a buffer in a variety of biological systems. Tris, MP Biomedicals is used for pH control of body fluids and in buffering systems for electrophoresis. It is ultra-pure, RNAse and DNAse-free, and relatively non-hygroscopic. It can be dried at 100°
67.73 € - 138.86 €
Chemical Identifiers
CAS | 77-86-1 |
---|---|
Molecular Formula | C4H11NO3 |
Molecular Weight (g/mol) | 121.136 |
InChI Key | LENZDBCJOHFCAS-UHFFFAOYSA-N |
Synonym | 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Trometamol, Tromethamine, Tris-[hydroxymethyl]amino-methane |
PubChem CID | 6503 |
ChEBI | CHEBI:9754 |
IUPAC Name | 2-amino-2-(hydroxymethyl)propane-1,3-diol |
SMILES | C(C(CO)(CO)N)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11473541
|
MP Biomedicals
02103133.2 |
250 g |
67.73 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11483541
|
MP Biomedicals
02103133.5 |
500 g |
138.86 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tris have been useful as buffers in a wide variety of biological systems. It is used in pH control in vitro and in vivo for body fluids and in buffering systems for electrophoresis applications.
- Used in assays used to characterize the activity and kinetics of the enzymes that catalyze SUMOylation of Small ubiquitin-like proteins (SUMO) and SUMO-dependent protein-protein interactions
- Used as a starting material for polymers, oxazolones (with carboxylic acids) and oxazolidines (with aldehydes)
- Does not precipitate calcium salts and is of value in maintaining solubility of manganese salts
- Can be used for the direct standardization of a strong acid solution; the equivalence point can be determined either potentiometrically or by use of a suitable indicator such as 3-(4-Dimethylamino-1-naphthylazo)-4-methoxybenzenesulfonic acid
Chemical Identifiers
77-86-1 | |
121.136 | |
2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Trometamol, Tromethamine, Tris-[hydroxymethyl]amino-methane | |
CHEBI:9754 | |
C(C(CO)(CO)N)O |
C4H11NO3 | |
LENZDBCJOHFCAS-UHFFFAOYSA-N | |
6503 | |
2-amino-2-(hydroxymethyl)propane-1,3-diol |
Specifications
77-86-1 | |
C4H11NO3 | |
Soluble in water (550mg/mL), ethylene glycol (79.1mg/mL), methanol (26mg/mL), anhydrous ethanol (14.6mg/mL), 95% ethanol (22.0mg/mL), DMF (14mg/mL), acetone (2mg/mL), ethyl acetate (0.5mg/mL), olive oil (0.4mg/mL), and chloroform (0.05mg/mL). | |
C(C(CO)(CO)N)O | |
121.136 | |
CHEBI:9754 | |
>99.9% |
Crystalline Powder | |
2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Trometamol, Tromethamine, Tris-[hydroxymethyl]amino-methane | |
LENZDBCJOHFCAS-UHFFFAOYSA-N | |
2-amino-2-(hydroxymethyl)propane-1,3-diol | |
6503 | |
121.14 | |
Ultrapure |
Safety and Handling
Recommended Storage : Store at Room Temperature (15–30°C). Store Desiccated.