missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tropolone, 98%, Thermo Scientific Chemicals
41.71 € - 440.19 €
Chemical Identifiers
CAS | 533-75-5 |
---|---|
Molecular Formula | C7H6O2 |
Molecular Weight (g/mol) | 122.12 |
MDL Number | MFCD00004158 |
InChI Key | MDYOLVRUBBJPFM-UHFFFAOYSA-N |
Synonym | tropolone, purpurocatechol, 2-hydroxycyclohepta-2,4,6-trienone, 2,4,6-cycloheptatrien-1-one, 2-hydroxy, 2-hydroxytropone, 2-hydroxy-2,4,6-cycloheptatrien-1-one, 2-hydroxy-2,4,6-cycloheptatrienone, unii-7l6dl16p1t, ccris 6609, tropomyosins |
PubChem CID | 10789 |
ChEBI | CHEBI:79966 |
IUPAC Name | 2-hydroxycyclohepta-2,4,6-trien-1-one |
SMILES | C1=CC=C(C(=O)C=C1)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10615962
|
Thermo Scientific Acros
140580010 |
1 g | Glass bottle |
41.71 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10533881
|
Thermo Scientific Acros
140580050 |
5 g | Glass bottle |
127.64 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10276913
|
Thermo Scientific Acros
140580250 |
25 g | Glass bottle |
440.19 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
533-75-5 | |
122.12 | |
MDYOLVRUBBJPFM-UHFFFAOYSA-N | |
10789 | |
2-hydroxycyclohepta-2,4,6-trien-1-one |
C7H6O2 | |
MFCD00004158 | |
tropolone, purpurocatechol, 2-hydroxycyclohepta-2,4,6-trienone, 2,4,6-cycloheptatrien-1-one, 2-hydroxy, 2-hydroxytropone, 2-hydroxy-2,4,6-cycloheptatrien-1-one, 2-hydroxy-2,4,6-cycloheptatrienone, unii-7l6dl16p1t, ccris 6609, tropomyosins | |
CHEBI:79966 | |
C1=CC=C(C(=O)C=C1)O |
Specifications
533-75-5 | |
Beige-Cream to White | |
>110°C | |
97.5% min. (GC) | |
MFCD00004158 | |
tropolone, purpurocatechol, 2-hydroxycyclohepta-2,4,6-trienone, 2,4,6-cycloheptatrien-1-one, 2-hydroxy, 2-hydroxytropone, 2-hydroxy-2,4,6-cycloheptatrien-1-one, 2-hydroxy-2,4,6-cycloheptatrienone, unii-7l6dl16p1t, ccris 6609, tropomyosins | |
MDYOLVRUBBJPFM-UHFFFAOYSA-N | |
2-hydroxycyclohepta-2,4,6-trien-1-one | |
10789 | |
122.12 | |
Crystalline Powder |
50.0°C to 54.0°C | |
115.0°C (12.0 mmHg) | |
Authentic | |
C7H6O2 | |
15, 9963 | |
Solubility in water: soluble in water. Other solubilities: soluble in ethyl ether, acetone, organic solvents | |
C1=CC=C(C(=O)C=C1)O | |
122.12 | |
CHEBI:79966 | |
98% | |
Tropolone |
Safety and Handling
missing translation for 'einecsNumber' : 208-577-2
Product Identifier
- Tropolone
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title