Learn More
Thermo Scientific™ TSG General Purpose Freezers - Crated

The TSG Series of general purpose freezers are built with the performance, security, and quality features you have come to expect from Thermo Scientific™.
6050.10 € - 7849.05 €
Specifications
Description | TSG Series General Purpose Laboratory Freezer |
---|---|
Temperature Range | -12°C to -30°C |
Temperature (Setpoint) | -20°C |
Door Style | Solid |
Cabinet Material | Steel |
Product Code | Brand | Capacity (Metric) | Plug Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Plug Type | Price | Quantity & Availability | ||||
17157758
![]() |
Thermo Scientific™
TSG700FESCW |
700 L | UK |
6050.10 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17117738
![]() |
Thermo Scientific™
TSG1500FESCW |
1500 L | UK |
7849.05 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Thermo Scientific™ TSG Series general purpose laboratory freezers are designed with your needs in mind. The TSG series is ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing reagents, media, proteins, and samples used daily in the lab.
TSG Series Freezers are designed with your laboratory in mind, combining the performance, security, and quality required for everyday laboratory applications.
- Wide range of sizes from solid door 400 L to 1500 L double door.
- Whisper quite operation < 50 dBA
- White painted interior and exterior
- Adaptive Defrost
- Temperature, door ajar, and power failure (with battery backup) alarms.
- Low heat output
- Rear access port
- Keyed Lock
- LED interior lighting
- Green hydrocarbon refrigerants
- Remote alarm contacts
Specifications
TSG Series General Purpose Laboratory Freezer | |
-20°C | |
Steel | |
30 kg | |
Yes | |
1 Rear Access Port | |
Adaptive Defrost | |
Alarm | |
230 V | |
R290 |
-12°C to -30°C | |
Solid | |
Painted Steel | |
CE | |
Digital Microprocessor | |
12 Months | |
Crated | |
Smart-Vue Pro Compatible | |
50 Hz |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.