missing translation for 'onlineSavingsMsg'
Learn More

Heidolph Instruments Tubing Set

Tubing Set for vacuum and water connections.

Brand:  Heidolph Instruments 591-35000-00

Product Code. 10188031

  • 223.00 € / Each
Estimated Shipment: 30-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Tubing set with 12 m piece including hose clamps.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclavable
39.4 ft.
Rotary evaporators
6.25 m Vacuum Tubing, 6.25 m Water Tubing, Hose Clamps
PTFE
12 m
Non-sterile
Vaccum Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Heidolph Instruments Tubing Set > Tubing Set; Includes: 6.25 m Vacuum Tubing, 6.25 m Water Tubing, Hose Clamps

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.