missing translation for 'onlineSavingsMsg'
Learn More

Tungsten(V) dichloride triethoxide, 10% w/v in ethanol, Thermo Scientific™

72.00 € - 162.00 €

Chemical Identifiers

CAS 45000-93-9
Molecular Formula C6H18Cl2O3W
Molecular Weight (g/mol) 392.947
MDL Number MFCD00210651
InChI Key AQDCOTYKQNGVLX-UHFFFAOYSA-L
Synonym tungsten v dichloride triethoxide w/v in ethanol
PubChem CID 86208842
IUPAC Name dichlorotungsten;ethanol
SMILES CCO.CCO.CCO.Cl[W]Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11343548
SDS View Documents
Thermo Scientific Alfa Aesar
039171.AC
25 mL N/A N/A N/A
11353548
SDS View Documents
Thermo Scientific Alfa Aesar
039171.AE
100 mL N/A N/A N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H225-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tungsten(V) dichloride triethoxide, 10% w/v in ethanol, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.