Learn More
Buerkle™ Turn 'n' Spray™ Spray Bottles with Overhead Valve
Spray in any direction with Turn'n' Spray's special technology. Buerkle™ Turn'n'Spray™ Spray Bottles with Overhead Valve spray uninterrupted at any angle and reaches hard-to-access areas.
11.90 € - 13.75 €
Specifications
Includes Overhead valve
Material Polyethylene, Polypropylene, Silicon
View More Specs
Products 3
Product Code Brand Capacity (Metric) Height (Metric) Neck ID (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Height (Metric) Neck ID (Metric) Price Quantity & Availability  
11991431
View Documents
Buerkle™
0309-0002
250 mL 220 mm 18 mm
11.90 €
Each
In Stock
to see stock.
Add to basket
11901441
View Documents
Buerkle™
0309-0005
500 mL 240 mm 18 mm
13.75 €
Each
In Stock
to see stock.
Edge
Add to basket
11911441
View Documents
Buerkle™
0309-0010
1000 mL 295 mm 17.5 mm
13.60 €
Each
In Stock
to see stock.
Edge
Add to basket
Description
Description
Turn'n'Spray features a special technology allowing you to spray in any direction and reach hard-to-access places easily and without straining.
  • Uninterrupted spraying at any angle
  • Parts in contact with contents made of PP, PE, silicon and stainless steel
  • Nozzle diameter 0.6mm, nozzle is continuously adjustable
  • Amount sprayed per stroke: 1.2mL ± 0.1mL
Specifications
Specifications
Overhead valve
Polyethylene, Polypropylene, Silicon