missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polysorbate 20, Fisher BioReagents™
Non-ionic detergent
Brand: Fisher BioReagents BP337-500
44.64 EUR valid until 2025-05-31
Use promo code "24127" to get your promotional price.
This item is not returnable.
View return policy
Description
Non-ionic detergent
This nonionic detergent is used in the isolation of water-soluble membrane proteins.
Specifications
Polysorbate 20 | |
9005-64-5 | |
100°C | |
6 | |
96 to 108 | |
0.25% max. | |
(C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 | |
3% max. | |
Polyoxyethylene-20-sorbitan Monolaurate, Polyoxyethylenesorbitan monolaurate | |
CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO | |
522.68 | |
1227.54 |
Amber | |
Viscous Liquid | |
1.100g/cm³ | |
500 mL | |
Pass Test | |
HDPE Bottle | |
400 mPa/s at 25°C | |
MFCD00165986 | |
HMFKFHLTUCJZJO-UHFFFAOYNA-N | |
2-[2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate | |
443314 |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Polysorbate 20, Fisher BioReagents™ > HDPE Bottle; 500 mL
Spot an opportunity for improvement?Share a Content Correction