missing translation for 'onlineSavingsMsg'
Learn More

Twelve component Organic multi-element standard 200μg/mL of each, Fisher chemical™

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

708.00 € - 766.00 €

Specifications

Chemical Name or Material Twelve component solution
Concentration 200μg/mL of each component
Packaging Ampoule
Type Organic Multi-component Standard Solution
UN Number 1230
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
12984275
View Documents
Spectracer
CM 1121
5 mL Ampoule
708.00 €
5mL
Please to purchase this item. Need a web account? Register with us today!
12994275
View Documents
Spectracer
CM 1122
10 mL Ampoule
766.00 €
10mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

Twelve component solution
Ampoule
1230
Liquid
200μg/mL of each component
Organic Multi-component Standard Solution
Methanol
Videos
Safety and Handling

Safety and Handling

H225 ; H301+H311 ; H370

P210;P241;P260;P280;P301+P310;P302+P352;P303+P361+P353;P312;P330;P361+P364;P370+P378;P501

Danger

missing translation for 'shelfLife' : 12 Months

Product Identifier
  • Standard solution 12 Components in methanol

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H370-Causes damage to organs.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P361 + P364-Take off immediately all contaminated clothing and wash it before reuse.
  • P370+P378-In case of fire: Use to extinguish.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications