Learn More
Twelve component Organic multi-element standard 200μg/mL of each, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Brand: Spectracer
Specifications
Twelve component solution | |
10 mL | |
1230 | |
Organic Multi-component Standard Solution | |
Methanol |
Ampoule | |
Liquid | |
200 μg/mL of each component | |
2-Chlorophenol, 2-Methylphenol, 2-Nitrophenol, 2,4-Dichlorophenol, 2,4-Dimethylphenol, 2,4,5-Trichlorophenol, 2,4,6-Trichlorophenol, 4-Chloro-3-methylphenol, 4-Methylphenol, 4-Nitrophenol, Pentachlorophenol, Phenol |
Safety and Handling
- Standard solution 12 Components in methanol
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 3
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H370-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P330-Rinse mouth.
- P361 + P364-Take off immediately all contaminated clothing and wash it before reuse.
- P370+P378-In case of fire: Use to extinguish.
- P501-Dispose of contents/container to .
Your input is important to us. Please complete this form to provide feedback related to the content on this product.